missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarboxylic Acid 96.0+%, TCI America™

Catalog No. p-7135643
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00811G 1 g
Q00815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00811G Supplier TCI America Supplier No. Q00811G
Only null left

Chemical Identifiers

CAS 879-65-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
MDL Number MFCD00012334
InChI Key UPUZGXILYFKSGE-UHFFFAOYSA-N
Synonym 2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
PubChem CID 96695
IUPAC Name quinoxaline-2-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC=CC2=N1

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 174.16
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Quinoxalinecarboxylic Acid