missing translation for 'onlineSavingsMsg'
Learn More

2-Quinolinecarbonitrile 98.0+%, TCI America™

Catalog No. Q00991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00991G 1 g
Q00995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00991G Supplier TCI America Supplier No. Q00991G

Chemical Identifiers

CAS 1436-43-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00134341
InChI Key WDXARTMCIRVMAE-UHFFFAOYSA-N
Synonym 2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0
PubChem CID 74031
IUPAC Name quinoline-2-carbonitrile
SMILES N#CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 160°C
Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Quinolinecarbonitrile