missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC64201DA

 View more versions of this product

Catalog No. CC64201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Pyrrolidin-1-yl-1, 3-thiazole-5-carboxylic acid
96.5
97%
C8H10N2O2S
1g
KPQGRYOIUITVHX-UHFFFAOYSA-N
2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid
24229715
97%
941716-91-2
100.0
Amber Glass Bottle
MFCD09879943
2-pyrrolidin-1-yl thiazole-5-carboxylic acid, 2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-pyrrolidinyl-1,3-thiazole-5-carboxylic acid
OC(=O)C1=CN=C(S1)N1CCCC1
198.24
198.25

Chemical Identifiers

941716-91-2
198.24
KPQGRYOIUITVHX-UHFFFAOYSA-N
24229715
OC(=O)C1=CN=C(S1)N1CCCC1
C8H10N2O2S
MFCD09879943
2-pyrrolidin-1-yl thiazole-5-carboxylic acid, 2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-pyrrolidinyl-1,3-thiazole-5-carboxylic acid
2-pyrrolidin-1-yl-1,3-thiazole-5-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions