missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrimidinylthiourea, 97+%, Thermo Scientific™

Catalog No. p-7032282
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3368103 1g
AAH3368109 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3368103 Supplier Thermo Scientific Chemicals Supplier No. H3368103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 31437-20-4
Molecular Formula C5H6N4S
Molecular Weight (g/mol) 154.191
MDL Number MFCD02662218
InChI Key MCCLFFQZNBEOAV-UHFFFAOYSA-N
Synonym 1-pyrimidin-2-yl thiourea, n-pyrimidin-2-ylthiourea, pyrimidin-2-yl-thiourea, pyrimidin-2-yl thiourea, n-2-pyrimidinyl thiourea, n-pyrimidin-2ylthiourea, 1-pyrimidin-2-ylthiourea, 1-pyrimidin-2-yl-thiourea, amino pyrimidin-2-ylamino methane-1-thione
PubChem CID 2760516
IUPAC Name pyrimidin-2-ylthiourea
SMILES C1=CN=C(N=C1)NC(=S)N

Specifications

Quantity 1g
Formula Weight 154.19
Percent Purity ≥97%
Chemical Name or Material 2-Pyrimidinylthiourea
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only