missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridylamidoxime 98.0+%, TCI America™
Supplier: TCI America P259025G
Specifications
| 2-Pyridylamidoxime | |
| 118°C | |
| C6H7N3O | |
| 25 g | |
| NJNUYQDYGLUAAG-WAYWQWQTSA-N | |
| (Z)-nitroso(1H-pyridin-2-ylidene)methanamine | |
| 5712435 | |
| ≥98.0% (HPLC,T) |
| 1772-01-6 | |
| White-Yellow | |
| MFCD00085159 | |
| n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine | |
| C1=CC(=C(N)N=O)NC=C1 | |
| 137.142 | |
| 137.14 | |
| Crystalline Powder |
Chemical Identifiers
| 1772-01-6 | |
| 137.142 | |
| NJNUYQDYGLUAAG-WAYWQWQTSA-N | |
| 5712435 | |
| C1=CC(=C(N)N=O)NC=C1 |
| C6H7N3O | |
| MFCD00085159 | |
| n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine | |
| (Z)-nitroso(1H-pyridin-2-ylidene)methanamine |
Safety and Handling
TSCA : No