Learn More
2-Pyridinemethanol, 98+%
CAS: 586-98-1 | C6H7NO | 109.128 g/mol
Supplier: Thermo Scientific Chemicals A1500522
Description
2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2-Pyridinemethanol | |
| -6°C to -5°C | |
| 112°C to 113°C (16 mmHg) | |
| C6H7NO | |
| MFCD00006348 | |
| 107849 | |
| 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol | |
| SHNUBALDGXWUJI-UHFFFAOYSA-N | |
| pyridin-2-ylmethanol | |
| 11474 | |
| ≥98% |
| 586-98-1 | |
| 1.131 | |
| 120°C (248°F) | |
| 1.544 | |
| 100 g | |
| Hygroscopic | |
| Fully miscible in water. | |
| C1=CC=NC(=C1)CO | |
| 109.128 | |
| 109.13 |
Chemical Identifiers
| 586-98-1 | |
| 109.128 | |
| SHNUBALDGXWUJI-UHFFFAOYSA-N | |
| 11474 | |
| C1=CC=NC(=C1)CO |
| C6H7NO | |
| MFCD00006348 | |
| 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol | |
| pyridin-2-ylmethanol |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 209-592-7
RTECSNumber : UT4689000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only