missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineethanol 98.0+%, TCI America™

Catalog No. p-7130438
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H024625ML 25 mL
H0246500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H024625ML Supplier TCI America Supplier No. H024625ML
Only null left

Chemical Identifiers

CAS 103-74-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00006364
InChI Key BXGYBSJAZFGIPX-UHFFFAOYSA-N
Synonym 2-pyridineethanol, 2-2-hydroxyethyl pyridine, 2-2-pyridyl ethanol, pyridine-2-ethanol, 2-pyridin-2-yl ethan-1-ol, 2-pyridin-2-yl ethanol, 2-pyridinethanol, 2-pyridylethanol, 2-pyridine ethanol, 2-beta-hydroxyethyl pyridine
PubChem CID 7675
IUPAC Name 2-pyridin-2-ylethanol
SMILES C1=CC=NC(=C1)CCO

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 mL
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Pyridineethanol
EINECSNumber (5)-0719
RTECSNumber UT2970450
TSCA Yes