missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinecarboxylic Acid Hydrazide 98.0+%, TCI America™

Catalog No. P042625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P042625G Supplier TCI America Supplier No. P042625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1452-63-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00059782
InChI Key BAQLNPIEFOYKNB-UHFFFAOYSA-N
Synonym 2-Picolinyl Hydrazide, Picolinic Acid Hydrazide
PubChem CID 255881
IUPAC Name pyridine-2-carbohydrazide
SMILES NNC(=O)C1=CC=CC=N1

Specifications

Melting Point 100°C
Color White
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyridinecarboxylic Acid Hydrazide