missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridinecarboxaldehyde 98.0+%, TCI America™
Supplier: TCI America P0425500G
Specifications
| 2-Pyridinecarboxaldehyde | |
| -21°C | |
| 181°C | |
| MFCD00006290 | |
| 2810 | |
| CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
| pyridine-2-carbaldehyde | |
| 14273 | |
| 107.11 | |
| Liquid |
| 1121-60-4 | |
| Yellow | |
| C6H5NO | |
| 500 g | |
| 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
| C1=CC=NC(=C1)C=O | |
| 107.112 | |
| CHEBI:73012 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1121-60-4 | |
| 107.112 | |
| CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
| 14273 | |
| pyridine-2-carbaldehyde |
| C6H5NO | |
| MFCD00006290 | |
| 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
| CHEBI:73012 | |
| C1=CC=NC(=C1)C=O |
Safety and Handling
EINECSNumber : (5)-6697
TSCA : Yes