missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. P042525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042525G 25 g
P0425100G 100 g
P0425500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P042525G Supplier TCI America Supplier No. P042525G

Chemical Identifiers

CAS 1121-60-4
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006290
InChI Key CSDSSGBPEUDDEE-UHFFFAOYSA-N
Synonym 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
PubChem CID 14273
ChEBI CHEBI:73012
IUPAC Name pyridine-2-carbaldehyde
SMILES C1=CC=NC(=C1)C=O

Specifications

Melting Point -21°C
Color Yellow
Boiling Point 181°C
Quantity 25 g
UN Number 2810
Formula Weight 107.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pyridinecarboxaldehyde
EINECSNumber (5)-6697
TSCA Yes