missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineacetonitrile 98.0+%, TCI America™

Catalog No. P101425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P101425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P101425G Supplier TCI America Supplier No. P101425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2739-97-1
Molecular Formula C7H6N2
Quantity 25 g
MDL Number MFCD00006346
UN Number 3439
Synonym 2-pyridylacetonitrile, 2-pyridineacetonitrile, pyridine-2-acetonitrile, 2-pyridin-2-yl acetonitrile, 2-2-pyridyl acetonitrile, 2-cyanomethyl pyridine, pyridine-acetonitrile, pyridyl-2-acetonitrile, pyridin-2-ylacetonitrile, pyridin-2-yl acetonitrile
InChI Key UKVQBONVSSLJBB-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)CC#N
IUPAC Name 2-pyridin-2-ylacetonitrile
Molecular Weight (g/mol) 118.139
PubChem CID 75959
Formula Weight 118.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Pyridineacetonitrile
Show More Show Less