missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineacetonitrile, 97%

Catalog No. AAA1308706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1308706 5 g
AAA1308714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1308706 Supplier Thermo Scientific Chemicals Supplier No. A1308706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2739-97-1 | C7H6N2 | 118.139 g/mol

2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 2739-97-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00006346
InChI Key UKVQBONVSSLJBB-UHFFFAOYSA-N
Synonym 2-pyridylacetonitrile, 2-pyridineacetonitrile, pyridine-2-acetonitrile, 2-pyridin-2-yl acetonitrile, 2-2-pyridyl acetonitrile, 2-cyanomethyl pyridine, pyridine-acetonitrile, pyridyl-2-acetonitrile, pyridin-2-ylacetonitrile, pyridin-2-yl acetonitrile
PubChem CID 75959
IUPAC Name 2-pyridin-2-ylacetonitrile
SMILES C1=CC=NC(=C1)CC#N

Specifications

Melting Point 23°C to 25°C
Density 1.059
Boiling Point 66°C to 68°C (1 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.525
Quantity 5 g
UN Number UN3439
Beilstein 111215
Solubility Information Miscible with water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 2-Pyridineacetonitrile
Show More Show Less
Hazard Category H227-H301-H310-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 220-364-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only