missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™

Catalog No. CC36901CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC36901CB 250mg Amber Glass Bottle
CC36901DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC36901CB Supplier Thermo Scientific Chemicals Supplier No. CC36901CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116247-03-1
Molecular Formula C9H8N2O2S
Molecular Weight (g/mol) 208.24
MDL Number MFCD02854711
InChI Key LJGAQGZEJDQDAU-UHFFFAOYNA-N
Synonym 2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl, 2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid, 3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine, 2-3-pyridyl-1,3-thiazoline-4-carboxylic acid, acmc-20dw5p, 4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole
PubChem CID 3130734
IUPAC Name 2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1CSC(=N1)C1=CN=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material 2-Pyrid-3-yl-4, 5-dihydro-1, 3-thiazole-4-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.