missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinemethanol 98.0+%, TCI America™

Catalog No. P21291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21291G Supplier TCI America Supplier No. P21291G
Only null left

Chemical Identifiers

CAS 6705-33-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01693938
InChI Key LFCWHDGQCWJKCG-UHFFFAOYSA-N
Synonym 2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol
PubChem CID 201734
IUPAC Name (pyrazin-2-yl)methanol
SMILES OCC1=CN=CC=N1

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Pyrazinemethanol
RTECSNumber UQ3625000
TSCA No
Recommended Storage Refrigerator