missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinecarbonyl chloride, Technical Grade, Thermo Scientific™

Catalog No. CC03402DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03402DA 1 g Amber Glass Bottle
CC03402CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03402DA Supplier Thermo Scientific Chemicals Supplier No. CC03402DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19847-10-0
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.542
InChI Key TXJKATOSKLUITR-UHFFFAOYSA-N
Synonym 2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g
PubChem CID 88279
IUPAC Name pyrazine-2-carbonyl chloride
SMILES C1=CN=C(C=N1)C(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 142.54
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Pyrazinecarbonyl chloride