missing translation for 'onlineSavingsMsg'
Learn More

2-Propylvaleryl Chloride 99.0+%, TCI America™

Catalog No. P08245ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08245ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08245ML Supplier TCI America Supplier No. P08245ML

Chemical Identifiers

CAS 2936-08-5
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.66
MDL Number MFCD00051446
InChI Key PITHYUDHKJKJNQ-UHFFFAOYSA-N
Synonym 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
PubChem CID 76240
IUPAC Name 2-propylpentanoyl chloride
SMILES CCCC(CCC)C(=O)Cl

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 3265
Formula Weight 162.66
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylvaleryl Chloride
EINECSNumber (2)-0632
TSCA No