missing translation for 'onlineSavingsMsg'
Learn More

2-Propylvaleric Acid 99.0+%, TCI America™

Catalog No. P082325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082325ML 25 mL
P0823100ML 100 mL
P0823500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P082325ML Supplier TCI America Supplier No. P082325ML

Chemical Identifiers

CAS 99-66-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00002672
InChI Key NIJJYAXOARWZEE-UHFFFAOYSA-N
Synonym valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
PubChem CID 3121
ChEBI CHEBI:39867
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(O)=O

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 144.21
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propylvaleric Acid
RTECSNumber YV7875000
TSCA No