missing translation for 'onlineSavingsMsg'
Learn More

2-Propylthiophene 98.0+%, TCI America™

Catalog No. p-7135572
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
P24515G 5 g
P245125G 25 g
2 options

Catalog No. P24515G

Supplier: TCI America P24515G

Only null left

Chemical Identifiers

CAS 1551-27-5
Molecular Formula C7H10S
Molecular Weight (g/mol) 126.217
MDL Number MFCD00014533
InChI Key BTXIJTYYMLCUHI-UHFFFAOYSA-N
Synonym 2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
PubChem CID 73771
IUPAC Name 2-propylthiophene
SMILES CCCC1=CC=CS1

Specifications

Boiling Point 160°C
Quantity 5 g
UN Number 1993
Formula Weight 126.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylthiophene