missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propylimidazole 95.0+%, TCI America™
$106.25 - $341.13
Chemical Identifiers
| CAS | 50995-95-4 |
|---|---|
| Molecular Formula | C6H10N2 |
| Molecular Weight (g/mol) | 110.16 |
| MDL Number | MFCD00059158 |
| InChI Key | MKBBSFGKFMQPPC-UHFFFAOYSA-N |
| PubChem CID | 162617 |
| IUPAC Name | 2-propyl-1H-imidazole |
| SMILES | CCCC1=NC=CN1 |
Chemical Identifiers
| 50995-95-4 | |
| 110.16 | |
| MKBBSFGKFMQPPC-UHFFFAOYSA-N | |
| 2-propyl-1H-imidazole |
| C6H10N2 | |
| MFCD00059158 | |
| 162617 | |
| CCCC1=NC=CN1 |
Specifications
| 50995-95-4 | |
| White-Yellow | |
| C6H10N2 | |
| 5 g | |
| CCCC1=NC=CN1 | |
| 110.16 | |
| 110.16 | |
| Crystalline Powder |
| 60°C | |
| 152°C | |
| MFCD00059158 | |
| MKBBSFGKFMQPPC-UHFFFAOYSA-N | |
| 2-propyl-1H-imidazole | |
| 162617 | |
| ≥95.0% (GC,T) | |
| 2-Propylimidazole |
Safety and Handling
EINECSNumber : (5)-0382
TSCA : Yes