missing translation for 'onlineSavingsMsg'
Learn More

2-Propylcyclohexanol (cis- and trans- mixture) 90.0+%, TCI America™

Catalog No. p-7134761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07705G 5 g
P077025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07705G Supplier TCI America Supplier No. P07705G

Chemical Identifiers

CAS 90676-25-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00019331
InChI Key VZBNUCDUQJCIDP-UHFFFAOYNA-N
PubChem CID 138614
IUPAC Name 2-propylcyclohexan-1-ol
SMILES CCCC1CCCCC1O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 142.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
TSCA No