missing translation for 'onlineSavingsMsg'
Learn More

2-Propyl-2-oxazoline 98.0+%, TCI America™

Catalog No. P17395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17395G 5 g
P17391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17395G Supplier TCI America Supplier No. P17395G
Only null left

Chemical Identifiers

CAS 4694-80-8
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
InChI Key GXCJLVVUIVSLOQ-UHFFFAOYSA-N
PubChem CID 78418
IUPAC Name 2-propyl-4,5-dihydro-1,3-oxazole
SMILES CCCC1=NCCO1

Specifications

Color Yellow
Boiling Point 147°C
Quantity 5 g
UN Number 1993
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propyl-2-oxazoline