missing translation for 'onlineSavingsMsg'
Learn More

2-Propoxyethylamine 97.0+%, TCI America™

Catalog No. A13255ML
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
A13255ML 5 mL
A132525ML 25 mL
2 options

Catalog No. A13255ML

Supplier: TCI America A13255ML

Only null left

Chemical Identifiers

CAS 42185-03-5
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00047955
InChI Key HMWXCSCBUXKXSA-UHFFFAOYSA-N
Synonym 2-Aminoethyl Propyl Ether, O-Propylethanolamine
PubChem CID 111878
IUPAC Name 2-propoxyethanamine
SMILES CCCOCCN

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 mL
UN Number 2733
Formula Weight 103.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propoxyethylamine