missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propionylthiophene 98.0+%, TCI America™
$102.52 - $102.52
Chemical Identifiers
| CAS | 13679-75-9 |
|---|---|
| Molecular Formula | C7H8OS |
| Molecular Weight (g/mol) | 140.2 |
| MDL Number | MFCD00005446 |
| InChI Key | MFPZQZZWAMAHOY-UHFFFAOYSA-N |
| Synonym | 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone |
| PubChem CID | 26179 |
| IUPAC Name | 1-thiophen-2-ylpropan-1-one |
| SMILES | CCC(=O)C1=CC=CS1 |
Chemical Identifiers
| 13679-75-9 | |
| 140.2 | |
| MFPZQZZWAMAHOY-UHFFFAOYSA-N | |
| 26179 | |
| CCC(=O)C1=CC=CS1 |
| C7H8OS | |
| MFCD00005446 | |
| 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone | |
| 1-thiophen-2-ylpropan-1-one |
Specifications
| 13679-75-9 | |
| C7H8OS | |
| 25 g | |
| MFPZQZZWAMAHOY-UHFFFAOYSA-N | |
| 1-thiophen-2-ylpropan-1-one | |
| 26179 | |
| ≥98.0% (GC) | |
| 2-Propionylthiophene |
| 228°C | |
| MFCD00005446 | |
| 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone | |
| CCC(=O)C1=CC=CS1 | |
| 140.2 | |
| 140.20 | |
| Liquid |
Safety and Handling
TSCA : No