missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylpyrrole, 99%

Catalog No. p-7027301
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2395403 1 g
AAB2395406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2395403 Supplier Thermo Scientific Chemicals Supplier No. B2395403
Only null left

CAS: 1073-26-3 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073-26-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD01696449
InChI Key AEJPPSRYXGEVDT-UHFFFAOYSA-N
Synonym 2-propionylpyrrole, 1-1h-pyrrol-2-yl propan-1-one, 2-pyrryl ethyl ketone, ethyl 2-pyrrolyl ketone, alpha-propionylpyrrole, 1-2-pyrrolyl-1-propanone, alpha-pyrryl ethyl ketone, 1-propanone, 1-pyrrol-2-yl, 1-1h-pyrrol-2-yl-1-propanone, unii-hgs11xf57f
PubChem CID 61260
IUPAC Name 1-(1H-pyrrol-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=CN1

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Formula Weight 123.15
Percent Purity 99%
Chemical Name or Material 2-Propionylpyrrole
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-026-7
RTECSNumber UC3805000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only