missing translation for 'onlineSavingsMsg'
Learn More

2-Propanethiol 96.0+%, TCI America™

Catalog No. p048925ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
P048925ML 25 mL
P0489500ML 500 mL
2 options

Catalog No. P048925ML

Supplier: TCI America P048925ML

Only null left

Chemical Identifiers

CAS 75-33-2
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004863
InChI Key KJRCEJOSASVSRA-UHFFFAOYSA-N
Synonym 2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
PubChem CID 6364
ChEBI CHEBI:8474
IUPAC Name propane-2-thiol
SMILES CC(C)S

Specifications

Melting Point -130°C
Color Colorless
Boiling Point 53°C
Quantity 25 mL
UN Number 2402
Formula Weight 76.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propanethiol
EINECSNumber (2)-0464
RTECSNumber TZ7302000
TSCA Yes