missing translation for 'onlineSavingsMsg'
Learn More

2-Propanethiol, 98%

Catalog No. p-187533
Change view
Click to view available options
Quantity:
50 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131440500 50 mL Glass bottle
AC131445000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131440500 Supplier Thermo Scientific Chemicals Supplier No. 131440500
Only null left

CAS: 75-33-2 | C3H8S | 76.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-33-2
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.15
MDL Number MFCD00004863
InChI Key KJRCEJOSASVSRA-UHFFFAOYSA-N
Synonym 2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
PubChem CID 6364
ChEBI CHEBI:8474
IUPAC Name propane-2-thiol
SMILES CC(C)S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -131°C
Density 0.8200g/mL
Boiling Point 57°C to 60°C
Flash Point -34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4245 to 1.4265
Linear Formula (CH3)2CHSH
Quantity 50 mL
Beilstein 01, 367
Specific Gravity 0.82
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohols,ethyl ether,hydrocarbons
Formula Weight 76.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Propanethiol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: Get medical advice/attention.
Keep away from heat/sparks/o
EINECSNumber 200-861-4
RTECSNumber TZ7302000
TSCA TSCA

RUO – Research Use Only