missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ 2-Point Touch Discriminator

Catalog No. S000883
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000883 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S000883 Supplier United Scientific™ Supplier No. TWOPOINT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prop 65

These tuning pipes create a long-lasting sound that fills an entire room.

This durable plastic tool is ideal for sensations testing. Two-point discrimination is a test used to assess if a person can identify two close points on a small area of skin. It has an additional point that allows you to switch from one to two-point simulations.

  • Made from ABS plastic
  • Easy to read scale
  • Single and double point stimulation
  • Ranges from 1 mm to 100 mm
  • Includes activity guide

Specifications

Grade 5+
Class Size 2 Students
Product Type Touch Discriminator Kit