missing translation for 'onlineSavingsMsg'
Learn More

2-(Pivalamido)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P20431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20431G Supplier TCI America Supplier No. P20431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146140-95-6
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD01114645
InChI Key MXRAJVMTCAUABO-UHFFFAOYSA-N
Synonym 2-tert-butylcarbonylamino phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivaloylamino phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivaloylaminobenzene boronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
PubChem CID 4193502
IUPAC Name [2-(2,2-dimethylpropanamido)phenyl]boronic acid
SMILES CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O

Specifications

Melting Point 271°C
Color White
Quantity 1 g
Formula Weight 221.06
Physical Form Crystalline Powder
Chemical Name or Material 2-(Pivalamido)phenylboronic Acid (contains varying amounts of Anhydride)