missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidineethanol 96.0+%, TCI America™

Catalog No. H073625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H073625G 25 g
H0736500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H073625G Supplier TCI America Supplier No. H073625G

Chemical Identifiers

CAS 1484-84-0
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00005989
InChI Key PTHDBHDZSMGHKF-UHFFFAOYSA-N
Synonym 2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol
PubChem CID 15144
IUPAC Name 2-piperidin-2-ylethanol
SMILES C1CCNC(C1)CCO

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 129.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Piperidineethanol
TSCA Yes
Recommended Storage Refrigerator