missing translation for 'onlineSavingsMsg'
Learn More

2-Picolylamine 98.0+%, TCI America™

Catalog No. p-7117276
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
A116125G 25 g
A1161250G 250 g
2 options

Catalog No. A116125G

Supplier: TCI America A116125G

Only null left

Chemical Identifiers

CAS 3731-51-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006360
InChI Key WOXFMYVTSLAQMO-UHFFFAOYSA-N
Synonym 2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
PubChem CID 19509
ChEBI CHEBI:81387
IUPAC Name 1-(pyridin-2-yl)methanamine
SMILES NCC1=CC=CC=N1

Specifications

Boiling Point 95°C
Quantity 25 g
UN Number 2735
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Picolylamine
EINECSNumber (5)-5596
RTECSNumber US1840000
TSCA Yes