missing translation for 'onlineSavingsMsg'
Learn More

2-Phthalimidoacetaldehyde Diethyl Acetal 95.0+%, TCI America™

Catalog No. P122925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P122925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P122925G Supplier TCI America Supplier No. P122925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78902-09-7
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.29
MDL Number MFCD00005901
InChI Key GEFXJJJQUSEHLV-UHFFFAOYSA-N
Synonym phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
PubChem CID 315286
IUPAC Name 2-(2,2-diethoxyethyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC

Specifications

Melting Point 72°C
Quantity 25 g
Formula Weight 263.29
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phthalimidoacetaldehyde Diethyl Acetal