missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water), TCI America™
$61.51 - $246.05
Chemical Identifiers
| CAS | 37971-36-1 |
|---|---|
| Molecular Formula | C7H11O9P |
| Molecular Weight (g/mol) | 270.13 |
| MDL Number | MFCD01940753 |
| InChI Key | SZHQPBJEOCHCKM-UHFFFAOYSA-N |
| Synonym | 3-Carboxy-3-phosphonohexanedioic Acid, PBTC |
| PubChem CID | 61973 |
| IUPAC Name | 2-phosphonobutane-1,2,4-tricarboxylic acid |
| SMILES | C(CC(CC(=O)O)(C(=O)O)P(=O)(O)O)C(=O)O |
Chemical Identifiers
| 37971-36-1 | |
| 270.13 | |
| SZHQPBJEOCHCKM-UHFFFAOYSA-N | |
| 61973 | |
| C(CC(CC(=O)O)(C(=O)O)P(=O)(O)O)C(=O)O |
| C7H11O9P | |
| MFCD01940753 | |
| 3-Carboxy-3-phosphonohexanedioic Acid, PBTC | |
| 2-phosphonobutane-1,2,4-tricarboxylic acid |
Specifications
| 37971-36-1 | |
| C7H11O9P | |
| 25 g | |
| 3-Carboxy-3-phosphonohexanedioic Acid, PBTC | |
| C(CC(CC(=O)O)(C(=O)O)P(=O)(O)O)C(=O)O | |
| 270.13 | |
| 270.13 | |
| 2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water) |
| Yellow | |
| MFCD01940753 | |
| 3265 | |
| SZHQPBJEOCHCKM-UHFFFAOYSA-N | |
| 2-phosphonobutane-1,2,4-tricarboxylic acid | |
| 61973 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2935
TSCA : Yes