missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water), TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 g |
| UN Number | 3265 |
| Formula Weight | 270.13 |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water) |
Safety and Handling
| EINECSNumber | (2)-2935 |
| TSCA | Yes |