missing translation for 'onlineSavingsMsg'
Learn More

2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water), TCI America™

Catalog No. P185725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P185725G 25 g
P1857500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P185725G Supplier TCI America Supplier No. P185725G
Only null left

Chemical Identifiers

CAS 37971-36-1
Molecular Formula C7H11O9P
Molecular Weight (g/mol) 270.13
MDL Number MFCD01940753
InChI Key SZHQPBJEOCHCKM-UHFFFAOYSA-N
Synonym 3-Carboxy-3-phosphonohexanedioic Acid, PBTC
PubChem CID 61973
IUPAC Name 2-phosphonobutane-1,2,4-tricarboxylic acid
SMILES C(CC(CC(=O)O)(C(=O)O)P(=O)(O)O)C(=O)O

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 270.13
Physical Form Liquid
Chemical Name or Material 2-Phosphonobutane-1,2,4-tricarboxylic Acid (ca. 50% in Water)
EINECSNumber (2)-2935
TSCA Yes