missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiazole-4-carboxylic acid, 97%

Catalog No. p-7036259
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54521MD 250 mg
AAH5452103 1 g
AAH5452106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54521MD Supplier Thermo Scientific Chemicals Supplier No. H54521MD

CAS: 7113-10-2 | C10H6NO2S | 204.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7113-10-2
Molecular Formula C10H6NO2S
Molecular Weight (g/mol) 204.22
MDL Number MFCD00141954
InChI Key IBUSLNJQKLZPNR-UHFFFAOYSA-M
Synonym 2-phenylthiazole-4-carboxylic acid, 2-phenyl-4-carboxythiazole, 4-thiazolecarboxylic acid, 2-phenyl, 2-phenyl-thiazole-4-carboxylic acid, 2-phenyl-4-thiazolecarboxylic acid, 2-phenyl-1,3-thiadiazole-4-carboxylic acid, pubchem19074, 4-carboxy-2-phenylthiazole, ksc237i8r, phenyl-1,3-thiazole-4-carboxylic acid
PubChem CID 138928
IUPAC Name 2-phenyl-1,3-thiazole-4-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC(=CS2)C(=O)O

Specifications

Melting Point 148°C to 152°C
Quantity 250 mg
Formula Weight 205.23
Percent Purity 97%
Chemical Name or Material 2-Phenylthiazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only