missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylquinoline 98.0+%, TCI America™

Catalog No. P20571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20571G 1 g
P20575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20571G Supplier TCI America Supplier No. P20571G

Chemical Identifiers

CAS 612-96-4
Molecular Formula C15H11N
Molecular Weight (g/mol) 205.26
MDL Number MFCD00011568
InChI Key FSEXLNMNADBYJU-UHFFFAOYSA-N
Synonym quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940
PubChem CID 71545
IUPAC Name 2-phenylquinoline
SMILES C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 363°C
Quantity 1 g
Formula Weight 205.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylquinoline