missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylquinoline-4-carboxylic acid, 99%

Catalog No. AAA1873414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1873414 25 g
AAA1873422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1873414 Supplier Thermo Scientific Chemicals Supplier No. A1873414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132-60-5 | C16H11NO2 | 249.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132-60-5
Molecular Formula C16H11NO2
Molecular Weight (g/mol) 249.269
MDL Number MFCD00006750
InChI Key YTRMTPPVNRALON-UHFFFAOYSA-N
Synonym cinchophen, tervalon, 2-phenyl-4-quinolinecarboxylic acid, cinchophene, cinconal, phenoquin, quinofen, quinophan, quinophen, atofan
PubChem CID 8593
IUPAC Name 2-phenylquinoline-4-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O

Specifications

Melting Point 212°C to 218°C
Odor Odorless
Quantity 25 g
Beilstein 192803
Sensitivity Light sensitive
Merck Index 14,2288
Solubility Information Insoluble in water
Formula Weight 249.27
Percent Purity 99%
Chemical Name or Material 2-Phenylquinoline-4-carboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-067-1
RTECSNumber GD4025000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only