missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66301DA

Quantity 1g

 View more versions of this product

Catalog No. CC66301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

122773-97-1
200.197
BOAIYSRFGWBZCF-UHFFFAOYSA-N
747389
C1=CC=C(C=C1)C2=NC=C(C=N2)C(=O)O
C11H8N2O2
MFCD00559612
2-phenyl-5-pyrimidinecarboxylic acid, 5-carboxy-2-phenylpyrimidine, 5-pyrimidinecarboxylic acid, 2-phenyl, 2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenyl, acmc-1c8my, 5-carboxy-2-phenyl-1,3-diazine, 2-phenyl pyrimidine-5-carboxylic acid
2-phenylpyrimidine-5-carboxylic acid

Specifications

2-Phenylpyrimidine-5-carboxylic acid
96.5
97%
C11H8N2O2
MFCD00559612
BOAIYSRFGWBZCF-UHFFFAOYSA-N
2-phenylpyrimidine-5-carboxylic acid
747389
97%
122773-97-1
100.0
Amber Glass Bottle
1g
2-phenyl-5-pyrimidinecarboxylic acid, 5-carboxy-2-phenylpyrimidine, 5-pyrimidinecarboxylic acid, 2-phenyl, 2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenyl, acmc-1c8my, 5-carboxy-2-phenyl-1,3-diazine, 2-phenyl pyrimidine-5-carboxylic acid
C1=CC=C(C=C1)C2=NC=C(C=N2)C(=O)O
200.197
200.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions