Learn More
2-Phenylpyridine, 98%
CAS: 1008-89-5 | C11H9N | 155.20 g/mol
Supplier: Thermo Scientific Chemicals L0786109
Description
2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.
Solubility
Fully miscible with ethanol, acetone, toluene, methylene chloride. Not miscible in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
| 2-Phenylpyridine | |
| -5°C | |
| 268°C to 270°C | |
| C11H9N | |
| MFCD00006280,MFCD31699954 | |
| 110445 | |
| Fully miscible with ethanol,acetone,toluene,methylene chloride. Not miscible in water. | |
| C1=CC=C(C=C1)C1=CC=CC=N1 | |
| 155.20 | |
| 155.2 |
| 1008-89-5 | |
| 1.086 | |
| >110°C (230°F) | |
| 1.624 | |
| 10 g | |
| pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine???? | |
| VQGHOUODWALEFC-UHFFFAOYSA-N | |
| 2-phenylpyridine | |
| 13887 | |
| 98% |
Chemical Identifiers
| 1008-89-5 | |
| 155.20 | |
| VQGHOUODWALEFC-UHFFFAOYSA-N | |
| 13887 | |
| C1=CC=C(C=C1)C1=CC=CC=N1 |
| C11H9N | |
| MFCD00006280,MFCD31699954 | |
| pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine???? | |
| 2-phenylpyridine |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-763-1
RTECSNumber : UT7140900
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only