missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropionitrile 90.0+%, TCI America™

Catalog No. p-7134815
Change view
Click to view available options
Quantity:
1 mL
10 mL
Catalog No. Quantity
P08601ML 1 mL
P086010ML 10 mL
2 options

Catalog No. P08601ML

Supplier: TCI America P08601ML

Only null left

Chemical Identifiers

CAS 1823-91-2
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001871
InChI Key NVAOLENBKNECGF-UHFFFAOYNA-N
Synonym 2-phenylpropionitrile, hydratroponitrile, alpha-methylphenylacetonitrile, alpha-methylbenzyl cyanide, 2-phenylpropiononitrile, benzeneacetonitrile, .alpha.-methyl, methylbenzyl cyanide, .alpha.-phenylpropionitrile, .alpha.-phenylethyl cyanide, .alpha.-methylbenzyl cyanide
PubChem CID 15761
IUPAC Name 2-phenylpropanenitrile
SMILES CC(C#N)C1=CC=CC=C1

Specifications

Boiling Point 230°C
Quantity 1 mL
UN Number 3276
Formula Weight 131.18
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylpropionitrile
EINECSNumber (3)-3119
TSCA Yes