missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Phenylpropionic acid, 98%

Catalog No. p-7043421
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0070906 5 g
AAL0070914 25 g
AAL0070922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0070906 Supplier Thermo Scientific Chemicals Supplier No. L0070906
Only null left

CAS: 492-37-5 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-37-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002650
InChI Key YPGCWEMNNLXISK-UHFFFAOYSA-N
Synonym 2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
PubChem CID 10296
ChEBI CHEBI:48526
IUPAC Name 2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point -5°C
Density 1.1
Boiling Point 264°C to 266°C
Flash Point 148°C (298°F)
Refractive Index 1.523
Quantity 5 g
Beilstein 1863558
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material (+/-)-2-Phenylpropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-752-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only