missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpiperazine, 96%

Catalog No. p-7029070
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2650206 5 g
AAH2650214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2650206 Supplier Thermo Scientific Chemicals Supplier No. H2650206

CAS: 5271-26-1 | C10H14N2 | 162.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5271-26-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD01871362
InChI Key RIMRLBGNCLMSNH-UHFFFAOYSA-N
Synonym 2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
PubChem CID 250673
IUPAC Name 2-phenylpiperazine
SMILES C1CNC(CN1)C2=CC=CC=C2

Specifications

Melting Point 83°C to 87°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 162.24
Percent Purity 96%
Chemical Name or Material 2-Phenylpiperazine
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only