missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpentane, tech. 85%

Catalog No. aab2011406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2011406 5 g
AAB2011403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2011406 Supplier Thermo Scientific Chemicals Supplier No. B2011406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2719-52-0 | C11H16 | 148.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2719-52-0
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00027145
InChI Key LTHAIAJHDPJXLG-UHFFFAOYNA-N
Synonym 2-phenylpentane, sec-pentylbenzene, benzene, sec-pentyl, sec-amylbenzene, benzene, 1-methylbutyl, 1-phenyl-1-methylbutane, s-amylbenzene, d-2-phenyl-pentane, sec-amyl benzene, methylbutyl benzene
PubChem CID 17627
IUPAC Name pentan-2-ylbenzene
SMILES CCCC(C)C1=CC=CC=C1

Specifications

Density 0.86
Quantity 5 g
Formula Weight 148.25
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Phenylpentane
EINECSNumber 220-324-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only