missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylimidazoline 98.0+%, TCI America™

Catalog No. P0685500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0685500G 500 g
P068525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0685500G Supplier TCI America Supplier No. P0685500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 936-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005180
InChI Key BKCCAYLNRIRKDJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
PubChem CID 13639
IUPAC Name 2-phenyl-4,5-dihydro-1H-imidazole
SMILES C1CN=C(N1)C1=CC=CC=C1

Specifications

Melting Point 100°C
Color White-Yellow
Boiling Point 181°C
Quantity 500 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylimidazoline
EINECSNumber (5)-3540
RTECSNumber NJ4395500
TSCA Yes