missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylglycinol 98.0+%, TCI America™

Catalog No. P17925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17925G 5 g
P179225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17925G Supplier TCI America Supplier No. P17925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7568-92-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00130145
InChI Key IJXJGQCXFSSHNL-UHFFFAOYNA-N
Synonym 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
PubChem CID 92466
IUPAC Name 2-amino-2-phenylethan-1-ol
SMILES NC(CO)C1=CC=CC=C1

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 106°C
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylglycinol
RTECSNumber SG7600000
TSCA No
Recommended Storage Refrigerator