missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine 98.0+%, TCI America™

Catalog No. P0085500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0085500ML 500 mL
P008525ML 25 mL
P0085100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0085500ML Supplier TCI America Supplier No. P0085500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-04-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008184
InChI Key BHHGXPLMPWCGHP-UHFFFAOYSA-N
Synonym phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
PubChem CID 1001
ChEBI CHEBI:18397
IUPAC Name 2-phenylethanamine
SMILES C1=CC=C(C=C1)CCN

Specifications

Color Yellow
Quantity 500 mL
UN Number 2735
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Phenylethylamine
EINECSNumber (3)-0338
RTECSNumber SG8750000
TSCA Yes