missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phényléthhylamine 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Quantity | 25 mL |
| UN Number | 2735 |
| Formula Weight | 121.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-Phenylethylamine |
Sécurité et manipulation
| EINECSNumber | (3)-0338 |
| RTECSNumber | SG8750000 |
| TSCA | Yes |