missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine Hydroiodide 98.0+%, TCI America™

Catalog No. p-7135458
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22131G 1 g
P22135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22131G Supplier TCI America Supplier No. P22131G
Only null left

Chemical Identifiers

CAS 151059-43-7
Molecular Formula C8H12IN
Molecular Weight (g/mol) 249.095
InChI Key UPHCENSIMPJEIS-UHFFFAOYSA-N
Synonym Phenethylamine Hydroiodide, 2-Phenylethylammonium Iodide, Phenethylammonium Iodide
PubChem CID 91972166
IUPAC Name 2-phenylethanamine;hydroiodide
SMILES C1=CC=C(C=C1)CCN.I

Specifications

Melting Point 259°C
Color White
Quantity 1 g
Formula Weight 249.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylethylamine Hydroiodide