missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine, 99%

Catalog No. AAA13402AU
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13402AU 1000 mL
AAA13402AD 50 mL
AAA13402AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13402AU Supplier Thermo Scientific Chemicals Supplier No. A13402AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-04-0 | C8H11N | 121.183 g/mol

Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).

Chemical Identifiers

CAS 64-04-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008184
InChI Key BHHGXPLMPWCGHP-UHFFFAOYSA-N
Synonym phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
PubChem CID 1001
ChEBI CHEBI:18397
IUPAC Name 2-phenylethanamine
SMILES C1=CC=C(C=C1)CCN

Specifications

Melting Point -60°C
Density 0.964
pH 11.5
Boiling Point 200°C to 202°C
Flash Point 81°C (178°F)
Odor Amine-like
Refractive Index 1.5335
Linear Formula C6H5(CH2)2NH2
Quantity 1000 mL
UN Number UN2922
Beilstein 507488
Sensitivity Air Sensitive
Merck Index 14,7225
Solubility Information It is soluble in water.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2-Phenylethylamine
Show More Show Less
Hazard Category H227-H301-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 200-574-4
RTECSNumber SG8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only