missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Phenylethyl)phosphonic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 138°C |
| Quantity | 200 mg |
| Formula Weight | 186.15 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (2-Phenylethyl)phosphonic Acid |
Safety and Handling
| TSCA | No |